siRNA Details
| Gene Symbol: LINCR-0003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120607.1 | 6 | 722 | |||
| GenBank mRNA | HY112357.1 | - | 2 | 415 | ||
| HY137151.1 | - | 2 | 181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINCR-0003| Gene Symbol: LINCR-0003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120607.1 | 6 | 722 | |||
| GenBank mRNA | HY112357.1 | - | 2 | 415 | ||
| HY137151.1 | - | 2 | 181 | |||