siRNA Details
| Gene Symbol: LINCR-0003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120607.1 | 6 | 1006 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000509844.1 | - | 2 | 513 | Chr. 5 - 57614389 - 57617431 [+] on Build GRCh38 | |

LINCR-0003
Genomic Map