siRNA Details
| Gene Symbol: LINC01470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109877.1 | 5 | 657 | |||
| GenBank mRNA | AK092848.1 | - | 4 | 673 | ||
| AK123816.1 | - | 5 | 657 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01470| Gene Symbol: LINC01470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109877.1 | 5 | 657 | |||
| GenBank mRNA | AK092848.1 | - | 4 | 673 | ||
| AK123816.1 | - | 5 | 657 | |||