siRNA Details
| Gene Symbol: LINC01586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120369.1 | 3 | 412 | |||
| NR_120370.1 | 3 | 402 | ||||
| GenBank mRNA | AK054749.1 | - | 3 | 412 | ||
| BX488539.1 | - | 3 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01586| Gene Symbol: LINC01586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120369.1 | 3 | 412 | |||
| NR_120370.1 | 3 | 402 | ||||
| GenBank mRNA | AK054749.1 | - | 3 | 412 | ||
| BX488539.1 | - | 3 | 406 | |||