siRNA Details
Gene Symbol: LINC01179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121676.1 | 6 | 1527 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000507838.1 | - | 6 | 1527 | Chr. 4 - 165684639 - 165762778 [+] on Build GRCh38 |
