siRNA Details
| Gene Symbol: LOC102723526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134271.1 | 2 | 481 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000503267.5 | - | 2 | 481 | Chr. 5 - 17444010 - 17445930 [+] on Build GRCh38 | |

LOC102723526
Genomic Map