siRNA Details
| Gene Symbol: LOC105377590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133944.1 | 2 | 382 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000512874.1 | - | 2 | 297 | Chr. 4 - 185471516 - 185472263 [+] on Build GRCh38 | |

LOC105377590
Genomic Map