siRNA Details
Gene Symbol: LOC105377590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133944.1 | 2 | 456 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000512874.1 | - | 2 | 371 | Chr. 4 - 185471516 - 185472263 [+] on Build GRCh38 |
