siRNA Details
| Gene Symbol: LOC101929109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134278.1 | 2 | 1340 | |||
| GenBank mRNA | BC043525.1 | - | 2 | 1340 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000511327.1 | - | 2 | 1341 | Chr. 5 - 76606608 - 76608970 [-] on Build GRCh38 | |

LOC101929109
Genomic Map