siRNA Details
Gene Symbol: LOC100507388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121617.1 | 4 | 1931 | |||
GenBank mRNA | AK027257.1 | - | 4 | 1932 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000508031.1 | - | 4 | 1931 | Chr. 4 - 74955974 - 74970362 [-] on Build GRCh38 |
