siRNA Details
| Gene Symbol: LOC100506406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104630.1 | 2 | 309 | |||
| GenBank mRNA | BX107768.1 | - | 2 | 288 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000503269.1 | - | 2 | 288 | Chr. 5 - 35938822 - 35939993 [+] on Build GRCh38 | |

LOC100506406
Genomic Map