siRNA Details
Gene Symbol: LINC01377 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104617.1 | 1 | 68 | |||
GenBank mRNA | HY010738.1 | - | 1 | 69 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000505396.2 | - | 1 | 68 | Chr. 5 - 3178095 - 3181232 [+] on Build GRCh38 |
