siRNA Details
| Gene Symbol: LINC01377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104617.1 | 1 | 197 | |||
| GenBank mRNA | HY010738.1 | - | 1 | 198 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000505396.2 | - | 2 | 197 | Chr. 5 - 3178095 - 3181232 [+] on Build GRCh38 | |

LINC01377
Genomic Map