siRNA Details
| Gene Symbol: CTD-2201I18.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109930.1 | 8 | 1320 | |||
| GenBank mRNA | BC047373.1 | - | 8 | 1309 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000514042.1 | - | 8 | 1309 | Chr. 5 - 80066349 - 80083654 [-] on Build GRCh38 | |

CTD-2201I18.1
Genomic Map