siRNA Details
Gene Symbol: CCDC37-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103787.1 | 2 | 620 | |||
GenBank mRNA | BC033989.2 | - | 2 | 583 | ||
BI827527.1 | - | 2 | 672 | |||
BX109262.1 | - | 2 | 246 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC37-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103787.1 | 2 | 620 | |||
GenBank mRNA | BC033989.2 | - | 2 | 583 | ||
BI827527.1 | - | 2 | 672 | |||
BX109262.1 | - | 2 | 246 |