siRNA Details
| Gene Symbol: LOC440117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033970.1 | 3 | 556 | |||
| GenBank mRNA | BC037858.2 | - | 3 | 512 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000507482.6 | - | 3 | 556 | Chr. 12 - 126869490 - 126874690 [-] on Build GRCh38 | |

LOC440117
Genomic Map