siRNA Details
| Gene Symbol: LOC100288866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103773.1 | 3 | 930 | |||
| GenBank mRNA | AK303671.1 | - | 3 | 930 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000514506.1 | - | 3 | 930 | Chr. 17 - 49505657 - 49574064 [-] on Build GRCh38 | |

LOC100288866
Genomic Map