siRNA Details
Gene Symbol: LINC00536 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046215.1 | 11 | 2127 | |||
GenBank mRNA | BC040336.1 | - | 11 | 2128 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000505156.2 | - | 11 | 2127 | Chr. 8 - 115950511 - 116325059 [-] on Build GRCh38 |
