siRNA Details
Gene Symbol: LOC284865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038460.1 | 3 | 857 | |||
GenBank mRNA | AK092552.1 | - | 3 | 857 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000506039.1 | - | 3 | 857 | Chr. 22 - 20198729 - 20204918 [-] on Build GRCh38 |
