siRNA Details
| Gene Symbol: BFSP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135278.1 | 5 | 701 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000515542.1 | - | 5 | 701 | Chr. 3 - 133429269 - 133455776 [-] on Build GRCh38 | |

BFSP2-AS1
Genomic Map