siRNA Details
| Gene Symbol: SLC7A11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038380.1 | 2 | 315 | |||
| GenBank mRNA | BC040999.1 | - | 2 | 292 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC7A11-AS1| Gene Symbol: SLC7A11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038380.1 | 2 | 315 | |||
| GenBank mRNA | BC040999.1 | - | 2 | 292 | ||