siRNA Details
| Gene Symbol: LOC255187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122044.1 | 3 | 759 | |||
| GenBank mRNA | BX106527.1 | - | 3 | 621 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000509139.1 | - | 3 | 621 | Chr. 5 - 149063455 - 149109787 [+] on Build GRCh38 | |

LOC255187
Genomic Map