siRNA Details
| Gene Symbol: CACNA1G-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038439.1 | 2 | 1407 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000505793.1 | - | 2 | 1407 | Chr. 17 - 50556207 - 50562108 [-] on Build GRCh38 | |

CACNA1G-AS1
Genomic Map