siRNA Details
| Gene Symbol: LOC100506142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104182.1 | 2 | 362 | |||
| GenBank mRNA | BC039392.1 | - | 2 | 334 | ||
| BC048424.1 | - | 2 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506142| Gene Symbol: LOC100506142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104182.1 | 2 | 362 | |||
| GenBank mRNA | BC039392.1 | - | 2 | 334 | ||
| BC048424.1 | - | 2 | 362 | |||