siRNA Details
Gene Symbol: LOC102546226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105021.1 | 5 | 569 | |||
GenBank mRNA | BC043237.1 | - | 5 | 574 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102546226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105021.1 | 5 | 569 | |||
GenBank mRNA | BC043237.1 | - | 5 | 574 |