siRNA Details
| Gene Symbol: LOC105377448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_133945.1 | 3 | 260 | |||
| GenBank mRNA | BX115310.1 | - | 3 | 260 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000505607.1 | - | 2 | 175 | Chr. 4 - 138819954 - 138821465 [-] on Build GRCh38 | |

LOC105377448
Genomic Map