siRNA Details
| Gene Symbol: FZD10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033834.1 | 2, 3 | 1464 | |||
| GenBank mRNA | AK123838.1 | - | 2, 3 | 1464 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000509760.1 | - | 2, 3 | 1464 | Chr. 12 - 130151593 - 130161678 [-] on Build GRCh38 | |

FZD10-AS1
Genomic Map