siRNA Details
Gene Symbol: LOC101927495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110189.1 | 4 | 1607 | |||
GenBank mRNA | BC040894.1 | - | 4 | 1607 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000506329.2 | - | 4 | 1607 | Chr. 11 - 61588494 - 61607550 [+] on Build GRCh38 |
