siRNA Details
| Gene Symbol: LOC101927495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110189.1 | 4 | 1845 | |||
| GenBank mRNA | BC040894.1 | - | 4 | 1845 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000506329.2 | - | 4 | 1845 | Chr. 11 - 61588494 - 61607550 [+] on Build GRCh38 | |

LOC101927495
Genomic Map