siRNA Details
| Gene Symbol: LOC101927359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125885.1 | 9 | 970 | |||
| GenBank mRNA | BC040219.1 | - | 9 | 967 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000505289.5 | - | 9 | 967 | Chr. 4 - 133093860 - 133149113 [-] on Build GRCh38 | |

LOC101927359
Genomic Map