siRNA Details
Gene Symbol: LINC01618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040106.1 | 5 | 635 | |||
GenBank mRNA | AL080225.1 | - | 5 | 635 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000514957.1 | - | 5 | 635 | Chr. 4 - 52789994 - 52815464 [+] on Build GRCh38 |
