siRNA Details
| Gene Symbol: MGC32805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_051996.1 | 6 | 633 | |||
| GenBank mRNA | BC029465.1 | - | 6 | 633 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC32805| Gene Symbol: MGC32805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_051996.1 | 6 | 633 | |||
| GenBank mRNA | BC029465.1 | - | 6 | 633 | ||