siRNA Details
| Gene Symbol: LINC00989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038826.1 | 4 | 1185 | |||
| GenBank mRNA | AK123913.1 | - | 4 | 1186 | ||
| BC047610.1 | - | 3 | 692 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00989| Gene Symbol: LINC00989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038826.1 | 4 | 1185 | |||
| GenBank mRNA | AK123913.1 | - | 4 | 1186 | ||
| BC047610.1 | - | 3 | 692 | |||