siRNA Details
Gene Symbol: LINC00989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038826.1 | 4 | 986 | |||
GenBank mRNA | AK123913.1 | - | 4 | 987 | ||
BC047610.1 | - | 3 | 493 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038826.1 | 4 | 986 | |||
GenBank mRNA | AK123913.1 | - | 4 | 987 | ||
BC047610.1 | - | 3 | 493 |