siRNA Details
| Gene Symbol: LOC101927124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110045.1 | 4 | 695 | |||
| GenBank mRNA | BC041327.1 | - | 4 | 700 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927124| Gene Symbol: LOC101927124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110045.1 | 4 | 695 | |||
| GenBank mRNA | BC041327.1 | - | 4 | 700 | ||