siRNA Details
| Gene Symbol: LOC101927124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110045.1 | 4 | 1116 | |||
| GenBank mRNA | BC041327.1 | - | 4 | 1121 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000502430.2 | - | 4 | 1116 | Chr. 14 - 31420758 - 31452883 [+] on Build GRCh38 | |

LOC101927124
Genomic Map