siRNA Details
| Gene Symbol: LOC101929034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104602.1 | 2 | 374 | |||
| GenBank mRNA | BX096909.1 | - | 2 | 325 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000509455.1 | - | 2 | 374 | Chr. 5 - 1855970 - 1856568 [-] on Build GRCh38 | |

LOC101929034
Genomic Map