siRNA Details
| Gene Symbol: HNCAT21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104609.1 | 2 | 374 | |||
| GenBank mRNA | BC036254.1 | - | 2 | 374 | ||
| BC044609.1 | - | 2 | 373 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000503877.1 | - | 2 | 373 | Chr. 5 - 118282575 - 118284782 [+] on Build GRCh38 | |

HNCAT21
Genomic Map