siRNA Details
Gene Symbol: HNCAT21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104609.1 | 2 | 848 | |||
GenBank mRNA | BC036254.1 | - | 2 | 848 | ||
BC044609.1 | - | 2 | 847 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000503877.1 | - | 2 | 847 | Chr. 5 - 118282575 - 118284782 [+] on Build GRCh38 |
