siRNA Details
| Gene Symbol: LOC339978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134657.1 | 2 | 637 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000508484.1 | - | 2 | 637 | Chr. 4 - 54603211 - 54607131 [-] on Build GRCh38 | |

LOC339978
Genomic Map