siRNA Details
| Gene Symbol: LOC101929227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132790.1 | 2 | 1267 | |||
| GenBank mRNA | CR936773.1 | - | 2 | 1267 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000505153.2 | - | 2 | 1267 | Chr. 11 - 120867933 - 120894797 [-] on Build GRCh38 | |

LOC101929227
Genomic Map