siRNA Details
Gene Symbol: LOC101929227 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132790.1 | 2 | 5874 | |||
GenBank mRNA | CR936773.1 | - | 2 | 5873 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000505153.2 | - | 2 | 5874 | Chr. 11 - 120867933 - 120894797 [-] on Build GRCh38 |
