siRNA Details
| Gene Symbol: LOC105374631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134266.1 | 2 | 287 | |||
| GenBank mRNA | BX089348.1 | - | 2 | 287 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000507950.1 | - | 2 | 287 | Chr. 5 - 5069192 - 5078311 [-] on Build GRCh38 | |

LOC105374631
Genomic Map