siRNA Details
Gene Symbol: LOC101927780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110416.1 | 1 | 58 | |||
GenBank mRNA | BX648502.1 | - | 1 | 60 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000508827.1 | - | 1 | 58 | Chr. 14 - 61556320 - 61570653 [-] on Build GRCh38 |
