siRNA Details
| Gene Symbol: LOC101927780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110416.1 | 4 | 399 | |||
| NR_110417.1 | 3 | 135 | ||||
| GenBank mRNA | BX648502.1 | - | 4 | 401 | ||
| HY039622.1 | - | 4 | 366 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927780| Gene Symbol: LOC101927780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110416.1 | 4 | 399 | |||
| NR_110417.1 | 3 | 135 | ||||
| GenBank mRNA | BX648502.1 | - | 4 | 401 | ||
| HY039622.1 | - | 4 | 366 | |||