siRNA Details
Gene Symbol: LINC01511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125810.1 | 1, 2 | 515 | |||
GenBank mRNA | BC034612.1 | - | 1, 2 | 507 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125810.1 | 1, 2 | 515 | |||
GenBank mRNA | BC034612.1 | - | 1, 2 | 507 |