siRNA Details
Gene Symbol: LINC00604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX115904.1 | - | 1 | 69 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000508437.1 | - | 1 | 69 | Chr. 5 - 40260657 - 40267657 [+] on Build GRCh38 |
