siRNA Details
| Gene Symbol: LOC100506272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110436.1 | 2, 3 | 167 | |||
| GenBank mRNA | BX108713.1 | - | 2, 3 | 168 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000507817.1 | - | 2, 3 | 167 | Chr. 4 - 187532878 - 187672641 [-] on Build GRCh38 | |

LOC100506272
Genomic Map