siRNA Details
| Gene Symbol: LINC01182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121681.1 | 4 | 1748 | |||
| GenBank mRNA | AK091889.1 | - | 4 | 1745 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000510907.5 | - | 4 | 1748 | Chr. 4 - 13655179 - 13931228 [+] on Build GRCh38 | |

LINC01182
Genomic Map