siRNA Details
| Gene Symbol: LINC00491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103753.1 | 3 | 889 | |||
| NR_103754.1 | 4 | 874 | ||||
| NR_103755.1 | 3 | 811 | ||||
| NR_103756.1 | 2 | 740 | ||||
| GenBank mRNA | AK092078.1 | - | 2 | 1031 | ||
| BG777293.1 | - | 2 | 694 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000510145.1 | - | 2 | 835 | Chr. 5 - 102609156 - 102671559 [-] on Build GRCh38 | |

LINC00491
Genomic Map